Skip to Content
Merck
All Photos(1)

Key Documents

C1-BIOC

Sigma-Aldrich

Collagenase Type I, Cls I

Synonym(s):

Collagen A

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$69.20
1 G
$500.00

$69.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$69.20
1 G
$500.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.78

$69.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

General description

Collagenase Type I (Cls I) is a tissue dissociation enzyme produced by Clostridium histolyticum. It digests large, intact triple helices of native collagen fibers. Collagenases belong to the matrix metalloproteinase (MMPs) family that digest collagens in the extracellular matrix (ECM). Collagens serve to be a major component of the extracellular matrix and in mammals. These structural proteins are triple helixes composed of three collagen polypeptides with high tensile strength.[1]
This product is identical to Biochrom GmbH part numbers C1-22 and C1-28. Enzyme blends of collagenase, clostripain, with tryptic and proteolytic activities from Clostridium histolyticum. This preparation is recommended for liver, lung, adipose, and adrenal tissue. Specific activity is 125 to 250 Mandl units per ml of powder substance.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tissue dissociation enzymes for adipose stromal vascular fraction cell isolation: a review
Lockhart RA, et al.
Journal of stem cell research & therapy (2015)

Questions

  1. Sehr geehrte Damen und Herren, in wie viel µl wird denn die Kollagenase Typ I (100mg) gelöst? Vielen Dank und liebe Grüße Verena

    1 answer
    1. This product is in powder form. The material is not tested for solubility, however, other collagenase enzymes currently offered are typically prepared at 1-2 mg/mL in TESCA buffer (containing 50 mM TES, 0.36 mM Calcium chloride, pH 7.4 at 37 C. The minimum activity of this product is 125 units per milligram. The activity is lot specific and reported in the Certificate of Analysis. Please see the link below to access the required Certificate:
      https://www.sigmaaldrich.com/search

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service