Skip to Content
Merck
All Photos(1)

Key Documents

BP729

Estradiol hemihydrate

British Pharmacopoeia (BP) Assay Standard

Synonym(s):

β-Estradiol hemihydrate, 1,3,5-Estratriene-3,17β-diol, 3,17β-Dihydroxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2 · 0.5H2O
CAS Number:
Molecular Weight:
281.39
Beilstein:
1914275
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

estradiol

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

O.C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O.C[C@]56CC[C@H]7[C@@H](CCc8cc(O)ccc78)[C@@H]5CC[C@@H]6O

InChI

1S/2C18H24O2.H2O/c2*1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20;/h2*3,5,10,14-17,19-20H,2,4,6-9H2,1H3;1H2/t2*14-,15-,16+,17+,18+;/m11./s1

InChI key

ZVVGLAMWAQMPDR-WVEWYJOQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Estradiol hemihydrate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Estradiol and Norethisterone Tablets
  • Estradiol and Norethisterone Acetate Tablets
  • Estradiol Tablets
  • Estradiol Vaginal Tablets
  • Estradiol Transdermal Patches
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Pictograms

    Health hazardEnvironment

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

    Storage Class Code

    6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    María Jesús López et al.
    Human reproduction (Oxford, England), 29(7), 1432-1437 (2014-05-02)
    How long is the individualized training and the stability of competence for the embryo transfer (ET) technique? The embryo transfer technique is easy-to-learn, hardly unlearned, and training should be individualized by monitoring with learning curve-cumulative summation (LC-CUSUM) curves. Like many
    Xiaohui Xia et al.
    Annals of translational medicine, 8(12), 762-762 (2020-07-11)
    To explore the effect of estrogen on human cerebral vascular smooth muscle cells (VSMCs) and to clarify the molecular mechanism of estrogen inhibition of VSMC proliferation, which could provide an important reference basis for the clinical treatment of hypertensive intracerebral
    Sadik Sahin et al.
    Archives of gynecology and obstetrics, 291(1), 185-191 (2014-08-01)
    The aim of this study was to compare the incidence of ectopic pregnancy in GnRH agonist triggered IVF cycles with intensive luteal support versus hCG triggered IVF cycles. This study was conducted as a retrospective cohort analysis of women who
    M Sousa et al.
    Zygote (Cambridge, England), 23(1), 145-157 (2013-09-03)
    Human oocyte dysmorphisms attain a large proportion of retrieved oocytes from assisted reproductive technology (ART) treatment cycles. Extracytoplasmic defects involve abnormal morphology of the zona pellucida (ZP), perivitelline space and first polar body. The aim of the present study was

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service