Skip to Content
Merck
All Photos(1)

Key Documents

BCR706

Pig kidney (CTC free)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$281.00

$281.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$281.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$281.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

storage temp.

−20°C

Analysis Note

For more information please see:
BCR706

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Antibiotics: Chlortetracycline

Matrix Group: Meat

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service