Skip to Content
Merck
All Photos(1)

Key Documents

BCR420

Waste mineral oil (PCB)

BCR®, certified reference material, low level

Synonym(s):

Mineral oil

Sign Into View Organizational & Contract Pricing

Select a Size

7.5 G
$810.00

$810.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
7.5 G
$810.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$810.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

neat

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR420

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service