Skip to Content
Merck
All Photos(1)

Key Documents

BCR024C

Titanium (O, N)

BCR®, certified reference material, cubes (0.2 g)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$369.00

$369.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$369.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$369.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

form

cubes (0.2 g)

manufacturer/tradename

JRC

format

matrix material

General description

Certified for the mass fractions of nitrogen and oxygen.

Analysis Note

For more information please see:
BCR024C

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service