Skip to Content
Merck
All Photos(1)

Key Documents

B3426

Millipore

Bile salts mixture

suitable for microbiology

Synonym(s):

Bile acids sodium salt, cholic acid-deoxycholic acid sodium salt mixture

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$371.00

$371.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$371.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

$371.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

packaging

pkg of 100 g

loss

≤6% loss on drying

color

yellow to brownish-yellow

pH

7±0.5

solubility

distilled water: freely soluble

application(s)

microbiology

storage temp.

10-30°C

General description

Bile salts mixture is a combination of an organic sodium salt with a conjugate of any of the bile acids along with either glycine or taurine. Bile salts are composed of four different kinds of bile acids - cholic, deoxycholic, chenodeoxycholic, and lithocholic acids. Bile salts are extracted under controlled conditions from purified fresh bile, to be used in bacteriological culture media, where it behaves as a selective inhibitory agent that inhibits Gram-positive bacteria and induced germination. It also inhibits the outgrowth of the germinated spores but does not interfere with the growth of Enterobacteriaceae.

Application

Bile salts mixture is recommended in diverse media like MacConkey Agar, SS Agar, and Violet Red Bile Agar. It increases the selectivity of the media to Enterobacteriaceae by inhibiting Gram-positive bacteria. It is used as a selective agent in culture media for the selective growth of intestinal bacteria like E. coli, other fecal coliforms, and fecal Streptococci. But it is also used as a selective agent in media for the detection of Yersinia enterocolitica, Aeromonas hydrophila, and Vibrio species.

Preparation Note

Extracted from purified, fresh bile.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chhatra B Kunwar et al.
Microbiology (Reading, England), 166(12), 1149-1159 (2020-11-19)
Colonization of the gastrointestinal tract by enterohaemorrhagic Escherichia coli (EHEC) is critically dependent on its ability to sense and respond to various microenvironments within the host. EHEC exposure to physiologically relevant levels of bile salts upregulates the two-component system, pmrAB
Crystal Gadishaw-Lue et al.
Infection and immunity, 89(2) (2020-11-25)
During passage through the human gastrointestinal tract, enterohemorrhagic Escherichia coli (EHEC) is exposed to membrane-damaging bile in the small intestine. We previously reported that EHEC treatment with a physiological bile salt mixture upregulates basRS, encoding a two-component system, and arnBCADTEF

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

  1. What us the bile salt composition of this mixture? Is this constant between batches?

    1 answer
    1. This product is pooled from a bovine source. The bile salt mixture is composed of approximately 50% each of sodium deoxycholate and sodium cholate. The exact composition is not tested. The manufacturing process is consistent, however, given the nature of the source, some variability between lots is expected. The degree of variability does not negatively impact microbial growth. Please see section 3 of the Safety Data Sheet:
      https://www.sigmaaldrich.com/sds/sial/b3426

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service