Skip to Content
Merck
All Photos(1)

Key Documents

B2759

Sigma-Aldrich

Bismarck Brown Y (G)

Powder

Synonym(s):

Basic Brown 1, Basic Brown G, GX, GXP, Excelsior Brown, Leather Brown, Manchester Brown, Phenylene Brown, Vesuvine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N8 · 2HCl
CAS Number:
Molecular Weight:
419.31
Colour Index Number:
21000
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

product name

Bismarck Brown Y (G),

form

powder

composition

Dye content, ~50%

SMILES string

Cl[H].Cl[H].Nc1ccc(\N=N\c2cccc(c2)\N=N\c3ccc(N)cc3N)c(N)c1

InChI

1S/C18H18N8.2ClH/c19-11-4-6-17(15(21)8-11)25-23-13-2-1-3-14(10-13)24-26-18-7-5-12(20)9-16(18)22;;/h1-10H,19-22H2;2*1H/b25-23+,26-24+;;

InChI key

MCZVRBLCRZWFJH-SPBSJSFYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Garde et al.
Archives of andrology, 39(1), 1-9 (1997-07-01)
A procedure is described for simultaneously evaluating of sperm viability and acrosomal status in fixed deer spermatozoa. This technique, which is a modification of the triple-stain technique (TST) developed for human sperm, utilizes trypan blue to identify the percentage of
[Case of poisoning by the dye Bismark Brown].
N M Ageeva et al.
Sudebno-meditsinskaia ekspertiza, 29(3), 59-59 (1986-07-01)
K Vasanth Kumar et al.
Journal of hazardous materials, 126(1-3), 198-201 (2005-07-28)
Equilibrium parameters for Bismarck brown onto rice husk were estimated by linear least square and a trial and error non-linear method using Freundlich, Langmuir and Redlich-Peterson isotherms. A comparison between linear and non-linear method of estimating the isotherm parameters was
An-Chong Chao et al.
Bioresource technology, 91(2), 157-162 (2003-11-01)
Chitosan (CTS) is a good adsorbent for dyes but lacks the ability to adsorb cationic dyes. In this study, chitosan was modified to possess the ability to adsorb cationic dyes from water. Four kinds of phenol derivatives: 4-hydroxybenzoic acid (BA)
J K Choi et al.
Analytical biochemistry, 236(1), 82-84 (1996-04-05)
A rapid and sensitive protein staining method employing a mixed dye technique has been developed. Solutions of Coomassie brilliant blue R-250 (CB, 0.2%) and Bismark brown R (BBR, 0.05%) were mixed in the volume ratio of 1:0.75 for staining (final

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service