Skip to Content
Merck
All Photos(1)

Documents

ARK2190

SAFC

N,N-Dimethylacetamide

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.21

vapor density

3 (vs air)

Quality Level

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

refractive index

n20/D 1.437 (lit.)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

suitability

suitable for manufacturing use

storage temp.

room temp

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Dumit et al.
Health physics, 119(6), 715-732 (2020-11-17)
The administration of chelation therapy to treat significant intakes of actinides, such as plutonium, affects the actinide's normal biokinetics. In particular, it enhances the actinide's rate of excretion, such that the standard biokinetic models cannot be applied directly to the
Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that
Fatma Kayaci et al.
Nanoscale, 6(17), 10224-10234 (2014-07-25)
Oxygen vacancies (V(O)s) in ZnO are well-known to enhance photocatalytic activity (PCA) despite various other intrinsic crystal defects. In this study, we aim to elucidate the effect of zinc interstitials (Zn(i)) and V(O)s on PCA, which has applied as well
Inês Farinha et al.
Carbohydrate polymers, 130, 455-464 (2015-06-17)
Purified chitin-glucan complex (CGCpure) was extracted from Komagataella pastoris biomass using a hot alkaline treatment, followed by neutralization and repeated washing with deionized water. The co-polymer thus obtained had a β-glucan:chitin molar ratio of 75:25 and low protein and inorganic
Naoki Aizawa et al.
Neurourology and urodynamics, 34(4), 368-374 (2014-02-18)
We measured single-unit mechanosensitive afferent activities (SAAs) during reflexic, rhythmic bladder contractions (RBCs), and examined whether L-arginine, an NO substrate, and mirabegron, a β3-adrenoceptor agonist, and oxybutynin, an antimuscarinic agent, can affect the SAAs in such condition. Twenty-nine female Sprague-Dawley

Articles

Find best practices for using Pellicon® Capsules in ADC processing as well as data from two studies using Pellicon® Capsules and Pellicon® cassettes in the ADC manufacturing process.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service