Skip to Content
Merck
All Photos(1)

Documents

ARK2046

SAFC

Lofepramine hydrochloride

Pharma Manufacturing

Synonym(s):

N-(4-Chlorobenzoylmethyl)-3-(10,11-dihydro-5h-dibenz[b,f]azepin-5-yl)-n-methylpropylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28Cl2N2O
CAS Number:
Molecular Weight:
455.42
MDL number:
UNSPSC Code:
51292923

Quality Level

SMILES string

Cl[H].CN(CCCN1c2ccccc2CCc3ccccc13)CC(=O)c4ccc(Cl)cc4

InChI

1S/C26H27ClN2O.ClH/c1-28(19-26(30)22-13-15-23(27)16-14-22)17-6-18-29-24-9-4-2-7-20(24)11-12-21-8-3-5-10-25(21)29;/h2-5,7-10,13-16H,6,11-12,17-19H2,1H3;1H

InChI key

ZWZIQPOLMDPIQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lofepramine is an antidepressant; serotonin and norepinephrine re-uptake inhibitor (SNRI).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service