Skip to Content
Merck
All Photos(2)

Documents

A2514

Sigma-Aldrich

Albumin from goat

≥96% (agarose gel electrophoresis)

Synonym(s):

Albumin powder, Goat albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.21

biological source

goat

Quality Level

Assay

≥96% (agarose gel electrophoresis)

form

powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is a globular protein. Serum albumin is a member of the multigene protein family. Human serum albumin (HSA) consists of three homologous domains (I-III) that form a heart-shaped structure. Each domain is a result of two subdomains called A and B. Bovine serum albumin (BSA) has a structure that is very similar to HSA as it has a heart-shaped structure that has three homologous domains (I-III) and each domain has three sub-domains.

Application

Albumin from goat has been used to verify the specificity of radioimmunoassay (RIA)-780 and 805.

Biochem/physiol Actions

Albumin is a highly soluble, multifunctional and major circulating plasma protein. It plays a role in physiological and pharmacological areas. Albumin participates in the transport of metals, cholesterol, hormones, bile pigments, drugs, and fatty acids. It regulates osmotic pressure and distributes fluids between various compartments. In addition, plasma albumin exhibits anti-oxidant effects to minimize oxidative stress. Albumin attributes to the major portion of total serum antioxidant properties.

Preparation Note

Fraction V Powder

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauricio Sendeski et al.
Radiology, 251(3), 697-704 (2009-04-16)
To determine whether a type of contrast medium (CM), iodixanol, modifies outer medullary descending vasa recta (DVR) vasoreactivity and nitric oxide (NO) production in isolated microperfused DVR. Animal handling conformed to the Animal Care Committee Guidelines of all participating institutions.
Abdullah Hussain et al.
Immunology, 128(2), 236-244 (2009-09-11)
Anti-proteinase 3 antibodies are implicated in the pathogenesis of small vessel vasculitis. These are primarily immunoglobulin G (IgG), with different subclasses predominating at different stages of disease. However, little is known of their respective roles in pathogenesis. We have previously
Claus G Madsen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 92, 1-7 (2015-02-11)
This study describes how protein release from polymer matrices correlate with simple measurements on the intrinsic viscosity of the polymer solutions used for casting the matrices and calculations of the solubility parameters of polymers and solvents used. Matrices of poly(dl-lactide-co-glycolide)
Nikolay N Brandt et al.
Journal of biomedical optics, 20(5), 051015-051015 (2014-12-06)
The analysis of the structure-function relationship is extremely important in the study of proteins. The importance of function-related motions of large parts or subglobules of protein molecules stimulates the spectroscopic study in the low-frequency (terahertz) domain. However, only tentative assignments
Dandan Luo et al.
Journal of controlled release : official journal of the Controlled Release Society, 220(Pt A), 484-494 (2015-11-19)
Drug bioavailability is a key consideration for drug delivery systems. When loaded with doxorubicin, liposomes containing 5 molar % porphyrin-phospholipid (HPPH liposomes) exhibited in vitro and in vivo serum stability that could be fine-tuned by varying the drug-to-lipid ratio. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service