Skip to Content
Merck
All Photos(1)

Key Documents

A0700000

Amobarbital sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Amobarbital sodium salt, 5-Ethyl-5-isoamylbarbituric acid, Amytal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H17N2O3Na
CAS Number:
Molecular Weight:
248.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

amobarbital

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na].CCC1(CCC(C)C)C(=O)NC(=O)NC1=O

InChI

1S/C11H18N2O3.Na.H/c1-4-11(6-5-7(2)3)8(14)12-10(16)13-9(11)15;;/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16);;

InChI key

URLVKOXOUJHSEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amobarbital sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jihua Fu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 48(6), 2409-2428 (2018-08-20)
Previously, we confirmed that liver-synthesized 5-HT rather than non-liver 5-HT, acting on the 5-HT2 receptor (5-HT2R), modulates lipid-induced excessive lipid synthesis (ELS). Here, we further revealed the effects of the hepatocellular 5-HT system in diabetes-related disorders. Studies were conducted in
Xiaoyan Zhu et al.
Frontiers in neuroscience, 14, 318-318 (2020-04-23)
Gossypol is a yellow polyphenolic compounds extracted from roots, stems and seeds of cotton plants. Excessive intake of gossypol induces severe pathological signs of toxicity in livestock and wildlife. Currently, gossypol has received widespread attention for its toxic effects on
Xuzhao Li et al.
Histochemistry and cell biology, 149(1), 61-73 (2017-10-05)
De novo synthesis of the nucleotide CTP is catalyzed by the essential pyrimidine biosynthesis enzyme CTP synthase (CTPs), which forms large-scale filamentous structures consisting of CTPs termed cytoophidia in prokaryotes and in eukaryotes. Recent studies have shown that cytoophidia are
Á Horváth et al.
Physiology international, 106(4), 347-354 (2020-01-04)
Proprioception plays an essential role in motor control and in psychological functioning: it is the basis of body schema and the feeling of body ownership. There are individual differences in the processing accuracy of proprioceptive stimuli. Although proprioceptive acuity plays
Jie-Yi Du et al.
Life sciences, 168, 28-37 (2016-01-15)
Palmitate, a common saturated free fatty acid, induces endothelial apoptosis in vitro in culture endothelial cells and in vivo in type 2 diabetes mellitus (T2DM) patients. The present study aimed to investigate whether Kv1.5 regulates palmitate-induced endothelial apoptosis and endothelial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service