Skip to Content
Merck
All Photos(1)

Key Documents

A0360000

Alprenolol hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(o-Allylphenoxy)-3-(isopropylamino)-2-propanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO2 · HCl
CAS Number:
Molecular Weight:
285.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

alprenolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CC(C)NCC(O)COc1ccccc1CC=C

InChI

1S/C15H23NO2.ClH/c1-4-7-13-8-5-6-9-15(13)18-11-14(17)10-16-12(2)3;/h4-6,8-9,12,14,16-17H,1,7,10-11H2,2-3H3;1H

InChI key

RRCPAXJDDNWJBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alprenolol hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matías E Díaz Merino et al.
Journal of chromatography. A, 1634, 461685-461685 (2020-11-20)
In this study, we describe the experimental variables influencing enantioseparation of twelve β-blockers when analyzed under polar-organic, reversed-phase and hydrophilic interaction liquid chromatography conditions on a column with immobilized amylose tris(3-chloro-5-methylphenylcarbamate) as chiral stationary phase. Regarding polar-organic mode, two component
Laura C Manella et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(48), 11605-11615 (2017-10-27)
Norepinephrine (NE) has been shown to influence sensory, and specifically olfactory processing at the behavioral and physiological levels, potentially by regulating signal-to-noise ratio (S/N). The present study is the first to look at NE modulation of olfactory bulb (OB) in
Pavel Sistik et al.
Biomedical chromatography : BMC, 30(2), 217-224 (2015-06-23)
A UPLC/MS/MS method with simple protein precipitation has been validated for the fast simultaneous analysis of agomelatine, asenapine, amisulpride, iloperidone, zotepine, melperone, ziprasidone, vilazodone, aripiprazole and its metabolite dehydro-aripiprazole in human serum. Alprenolol was applied as an internal standard. A
Sara Jamshidi et al.
Journal of separation science, 42(3), 698-705 (2018-11-19)
In this work, a novel, efficient, and green sorbent, SiO2 @Fe3 O4 has been created and functionalized with 1-butyl-3-methylimidazolium hexafluorophosphate as an ionic liquid. This sorbent was applied for microextraction of four beta blockers, propranolol, metoprolol, atenolol, and alprenolol with
Moataz Dowaidar et al.
Scientific reports, 7(1), 12635-12635 (2017-10-05)
Cell-penetrating peptides (CPPs) uptake mechanism is still in need of more clarification to have a better understanding of their action in the mediation of oligonucleotide transfection. In this study, the effect on early events (1 h treatment) in transfection by PepFect14

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service