95440
2-Ethyl-5-phenylisoxazolium-3′-sulfonate
purum, ≥97.0% (T)
Synonym(s):
Woodwards reagent K
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H11NO4S
CAS Number:
Molecular Weight:
253.27
Beilstein:
4149224
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Recommended Products
grade
purum
Assay
≥97.0% (T)
mp
216-219 °C (dec.)
SMILES string
CC[n+]1ccc(o1)-c2cccc(c2)S([O-])(=O)=O
InChI
1S/C11H11NO4S/c1-2-12-7-6-11(16-12)9-4-3-5-10(8-9)17(13,14)15/h3-8H,2H2,1H3
InChI key
MWOOKDULMBMMPN-UHFFFAOYSA-N
Other Notes
Modification of carboxylic groups in enzymes; Coupling reagent for peptide synthesis
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
V.L. Boyd et al.
Tetrahedron Letters, 31, 3849-3849 (1990)
A A Komissarov et al.
The Journal of biological chemistry, 270(17), 10050-10055 (1995-04-28)
Woodward's reagent K (WRK) completely inactivated Escherichia coli uridine phosphorylase by reversible binding in the active site (Ki = 0.07 mM) with subsequent modification of a carboxyl (k2 = 1.2 min-1). Neither substrate alone protected uridine phosphorylase from inactivation. The
G Feller et al.
The Journal of biological chemistry, 271(39), 23836-23841 (1996-09-27)
Chloride is the allosteric effector of vertebrate pancreatic and salivary alpha-amylases and of the bacterial alpha-amylase from Alteromonas haloplanctis. Activation experiments of A. haloplanctis alpha-amylase by several monovalent anions show that a negative charge, not restricted to that of Cl-
S R Rao et al.
Indian journal of biochemistry & biophysics, 34(3), 253-258 (1997-06-01)
Maize leaf NADP-malic enzyme was rapidly inactivated by micromolar concentrations of Woodward's reagent K (WRK). The inactivation followed pseudo-first order reaction kinetics. The order of reaction with respect to WRK was 1, suggesting that inactivation was a consequence of the
M S Saini et al.
Biochimica et biophysica acta, 568(2), 370-376 (1979-06-06)
Treatment of homogenous human prostatic acid phosphatase (orthophosphoric-monoester phosphohydrolase (acid optimum), EC 3.1.3.2) with low concentrations of Woodward's reagent K (N-ethyl-5-phenylisoxazolium-3'-sulfonate) leads to a rapid loss of enzymic activity. The rate of inactivation of the enzyme is reduced in the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service