Skip to Content
Merck
All Photos(1)

Key Documents

95343

Supelco

(±)-1-Amino-2-propanol

analytical standard

Synonym(s):

(±)-Isopropanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$104.00

$104.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$104.00

About This Item

Linear Formula:
CH3CH(OH)CH2NH2
CAS Number:
Molecular Weight:
75.11
Beilstein:
605275
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$104.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

2.6 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98.0% (GC)

optical purity

enantiomeric ratio: ~50:50 (racem)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.446-1.450
n20/D 1.4478 (lit.)

bp

160 °C (lit.)

mp

−2 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(O)CN

InChI

1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3

InChI key

HXKKHQJGJAFBHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J A Redondo et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 90, 38-43 (2014-12-03)
In this work, the cationic monomer N-ethyl pyrrolidine methacrylamide (EPA) was copolymerized with the neutral monomer N-hydroxypropyl methacrylamide (HPMA) at different molar ratios obtaining linear random copolymers that were characterized and evaluated in vitro as non-viral gene carriers using murine
Tomáš Etrych et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 58, 1-12 (2014-03-19)
In this study, we describe the synthesis, physico-chemical characterisation and results of the in vitro and in vivo evaluation of the biological behaviour of N-(2-hydroxypropyl)methacrylamide-based (HPMA) copolymer conjugates bearing doxorubicin (DOX) partly bound via a pH-sensitive hydrazone and partly via

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service