Skip to Content
Merck
All Photos(1)

Key Documents

94896

Sigma-Aldrich

1-Bis(4-fluorophenyl)methyl piperazine

purum, ≥97.0% (GC)

Synonym(s):

1-(4,4′-Difluorobenzhydryl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18F2N2
CAS Number:
Molecular Weight:
288.34
Beilstein:
620754
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

mp

88-92 °C (lit.)
88-92 °C

SMILES string

Fc1ccc(cc1)C(N2CCNCC2)c3ccc(F)cc3

InChI

1S/C17H18F2N2/c18-15-5-1-13(2-6-15)17(21-11-9-20-10-12-21)14-3-7-16(19)8-4-14/h1-8,17,20H,9-12H2

InChI key

TTXIFFYPVGWLSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Yamanaka et al.
Archives of toxicology, 69(6), 391-396 (1995-01-01)
Toxic effects of a detriazinyl metabolite of almitrine (DTMA) were evaluated in rats and on cultured rat macrophages. In rats daily treated with DTMA for 16 weeks, spastic gaits with heel-lifting appeared, and lamellated and/or crystalloid bodies formed in sensory
S Kariya et al.
Research communications in chemical pathology and pharmacology, 78(1), 85-95 (1992-10-01)
The oxidative metabolism of flunarizine [1-[bis(4-fluorophenyl)-methyl]-4-(3-phenyl-2-propenyl)piperazine, FZ] to 1-[bis-(4-fluorophenyl)methyl]piperazine (M-1), 1-[bis(4-fluorophenyl)methyl]-4-[3-(4'-hydroxyphenyl)-2- propenyl]piperazine (M-2) and 4,4'-difluorobenzophenone (M-3) has been studied in liver microsomes of Wistar and Dark Agouti (DA) rats. Kinetic analysis demonstrated a sex difference (male > female) in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service