Skip to Content
Merck
All Photos(1)

Key Documents

94234

Supelco

Alkane Standard Mixture for performance tests of GC-systems

analytical standard, C10; C20 - C40 (all even), 50 mg/l each

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$121.00
10 ML
$468.00

$121.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

Select a Size

Change View
2 ML
$121.00
10 ML
$468.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$121.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.388

density

0.684 g/mL at 20 °C

application(s)

environmental
petroleum

format

multi-component solution

Application

Alkane standard mixture has been used as a standard for the determination of long-chain hydrocarbons in the cuticular extracts of fruit flies and worker bees on etched silver foils as substrate.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

contains C10 and all even n-alkanes from C20 to C40, 50 mg/l in n-heptane each

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laser desorption/ionization mass spectrometry of lipids using etched silver substrates
Schnapp A, et al.
Methods, 104, 194-203 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service