940089
RuPhos Pd G2 Chembeads
Synonym(s):
2nd Generation RuPhos Precatalyst ChemBeads, Chloro(2-dicyclohexylphosphino-2′,6′-diisopropoxy-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II) ChemBeads, RuPhos-Pd-G2 ChemBeads
About This Item
Recommended Products
form
solid
Quality Level
composition
, 4-6 wt. % (loading)
reaction suitability
reagent type: catalyst
SMILES string
CC(OC(C=CC=C1OC(C)C)=C1C(C=CC=C2)=C2P(C3CCCCC3)C4CCCCC4)C.Cl[Pd]5C6=CC=CC=C6C7=CC=CC=C7N5
InChI
InChI=1S/C30H43O2P.C12H10N.ClH.Pd/c1-22(2)31-27-19-13-20-28(32-23(3)4)30(27)26-18-11-12-21-29(26)33(24-14-7-5-8-15-24)25-16-9-6-10-17-25;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h11-13,18-25H,5-10,14-17H2,1-4H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1
InChI key
VPDRBFBKNSFRSS-UHFFFAOYSA-M
General description
Application
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For general uses, product is also available in powdered form (753246)
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service