Skip to Content
Merck
All Photos(1)

Documents

93909

Supelco

1,3-Dimethylamylamine

analytical standard

Synonym(s):

1,3-Dimethylpentanamine, 1,3-Dimethylpentylamin, 4-Methyl-2-hexanamine, Methylhexanamine, NSC 1106

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17N
CAS Number:
Molecular Weight:
115.22
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.417-1.419

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)
veterinary

format

neat

InChI

1S/C7H17N/c1-4-6(2)5-7(3)8/h6-7H,4-5,8H2,1-3H3

InChI key

YAHRDLICUYEDAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dimethylamylamine is a neutral stimulant, which is widely used in pre-workout products as a dietary supplement.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.0 °F

Flash Point(C)

43.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 3?Dimethylamylamine (DMAA) in supplements and geranium products: natural or synthetic?
Zhang Y, et al.
Drug Testing and Analysis, 4, 986-990 (2012)
Nadali Ramezani et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 251, 119415-119415 (2021-01-27)
1,3-Dimethylpentylamine (Geranamine) with a similar structure to amphetamine has been used as an athletic performance promoter (doping agent) and also as an indirect sympathomimetic drug to synthesize of 1,3-dimethyl pentyl glycine (13DMPG). Thereafter, two new anticancer platinum complexes as [Pt(DACH)(13DMPG)]NO3
Hemodynamic and hematologic profile of healthy adults ingesting dietary supplements containing 1, 3-dimethylamylamine and caffeine
Farney.MT, et al.
Nutrition and metabolic insights, 5, 1-1 (2012)
Anna Rickli et al.
European journal of pharmacology, 859, 172515-172515 (2019-07-03)
Preworkout supplements ("boosters") are used to enhance physical and mental performance during workouts. These products may contain various chemical substances with undefined pharmacological activity. We investigated whether substances that are contained in commercially available athletic multiple-ingredient preworkout supplements exert amphetamine-type
J S Li et al.
Analytical chemistry insights, 7, 47-58 (2012-08-24)
A sensitive and reliable method of liquid chromatography-electrospray ionization/tandem mass spectrometry (LC-ESI/MS/ MS) was developed and validated for determining 1,3-dimethylamylamine (1,3-DMAA) and 1,4-dimethylamylamine (1,4-DMAA) in geranium plants (Pelargonium graveolens). The sample was extracted with 0.5 M HCl and purified by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service