Skip to Content
Merck
All Photos(2)

Key Documents

93492

Millipore

Wheat Peptone for Biotechnology and Microbiology

Enzymatically Digested, for Biotechnology and Microbiology

Synonym(s):

Peptone from wheat

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$155.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$155.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

$155.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Product Name

Wheat Peptone, suitable for microbiology

biological source

wheat

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥2%
total nitrogen (N), ≥11%

impurities

<24% ash

loss

<6% loss on drying

pH

6.8±0.5 (2% in H2O)

solubility

H2O: 2%, clear, light yellow to brown

application(s)

food and beverages
microbiology

suitability

Enterobacter spp.
Lactobacillus spp.
Listeria spp.

General description

Wheat Peptone is obtained by enzymatic hydrolysis of wheat proteins. This peptone favors the growth of Enterobacteria, and Listeria. It is also suited for the culture of Lactobacillus. By nature of its high protein content and relatively clear aspect. Wheat Peptone is particualrly well suited for broth cultures and applications where transparency is essential.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debra Rossouw et al.
Applied and environmental microbiology, 75(20), 6600-6612 (2009-08-25)
Commercial wine yeast strains of the species Saccharomyces cerevisiae have been selected to satisfy many different, and sometimes highly specific, oenological requirements. As a consequence, more than 200 different strains with significantly diverging phenotypic traits are produced globally. This genetic

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service