92277
Trimethylamine N-oxide dihydrate
purum, ≥99.0% (NT)
Synonym(s):
TMANO
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
purum
Quality Level
Assay
≥99.0% (NT)
form
crystals
reaction suitability
reagent type: oxidant
mp
95-99 °C (lit.)
95-99 °C
SMILES string
O.O.C[N+](C)(C)[O-]
InChI
1S/C3H9NO.2H2O/c1-4(2,3)5;;/h1-3H3;2*1H2
InChI key
PGFPZGKEDZGJQZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Trimethylamine N-oxide (TMAO) is an amphiphilic osmolyte that can counteract the denaturing effects of urea, pressure, and ice and stabilize the proteins.
Application
Reactant for:
- C-H bond cleavage
- Oxidation reactions (oxidant)
- Decarbonylating agent for solvent-free reactions
Trimethylamine N-oxide dihydrate can react with transition-metal carbonyl cluster compounds and activate CO displacement by converting CO ligands into carbon dioxide.
Other Notes
Oxidant for the catalytic OsO4 cis-hydroxylation of hindered olefins; preparation of the unstable, crystalline anhydrous compound
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tetrahedron Letters, 21, 449-449 (1980)
Counteraction of urea by trimethylamine N-oxide is due to direct interaction.
Biophysical Journal, 97(9), 2559-2566 (2009)
Trimethylamine N-oxide promoted reactions of manganese, molybdenum and tungsten carbonyl complexes.
Journal of Organometallic Chemistry, 173(1), 71-76 (1979)
Tetrahedron Letters, 27, 3961-3961 (1986)
Cluster synthesis. 10. Reaction of Os3 (CO) 10 (. mu. 3-S) with trimethylamine N-oxide dihydrate. Syntheses and structural characterizations of Os3 (CO) 8 (NMe3)(. mu.-OH)(. mu. 3-S)(. mu.-H) and the six-atom-chain cluster Os6 (CO) 18 (. mu.-OH)(. mu. 4-S)(. mu. 3-S)(. mu.-H).
Inorganic Chemistry, 25(8), 1122-1127 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service