Skip to Content
Merck
All Photos(1)

Key Documents

91904

Supelco

Violaxanthin

analytical standard

Synonym(s):

(3S,3′S,5R,5′R,6S,6′S)-5,6:5′,6′-Diepoxy-5,5′,6,6′-tetrahydro-β,β-carotene-3,3′-diol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,550.00

$1,550.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$1,550.00

About This Item

Empirical Formula (Hill Notation):
C40H56O4
CAS Number:
Molecular Weight:
600.87
Beilstein:
101269
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.24

$1,550.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−70°C

SMILES string

CC(/C=C/[C@@]12[C@](C)(O2)C[C@@H](O)CC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(\C=C\[C@]34C(C)(C)C[C@H](O)C[C@]3(O4)C)C

InChI

1S/C40H56O4/c1-29(17-13-19-31(3)21-23-39-35(5,6)25-33(41)27-37(39,9)43-39)15-11-12-16-30(2)18-14-20-32(4)22-24-40-36(7,8)26-34(42)28-38(40,10)44-40/h11-24,33-34,41-42H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+/t33-,34-,37+,38+,39-,40-/m0/s1

InChI key

SZCBXWMUOPQSOX-WVJDLNGLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Violaxanthins are natural pigmented constituents or carotenoids present in most fruits and vegetables.[1]

Application

Violaxanthin may be used as a reference standard for the determination of the analyte in fruits and vegetables by high performance liquid chromatography with photodiode array detection.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the carotenoid content in selected vegetables and fruit by HPLC and photodiode array detection.
Muller H.
European Food Research and Technology, 204(2), 88- 94 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service