Skip to Content
Merck
All Photos(3)

Key Documents

918598

Sigma-Aldrich

CX234

Umicore

Synonym(s):

Chloro[1,3-bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](1-tert-butyl-1H-inden-1-yl)palladium(II), [(IPent)Pd(1-tBu-Ind)Cl]

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$511.00

$511.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$511.00

About This Item

Empirical Formula (Hill Notation):
C48H67ClN2Pd
Molecular Weight:
813.93
UNSPSC Code:
12352101
NACRES:
NA.22

$511.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Quality Level

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

Application

Umicore CX234 is a homogeneous catalyst useful for cross-coupling reactions with arenes, Buchwald-Hartwig aminaton reactions, and Suzuki-Miyaura cross-couplings.

System Requirements

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service