Skip to Content
Merck
All Photos(2)

Key Documents

917990

Sigma-Aldrich

Hexahydro-1,3-bis(4-methylphenyl)-5-phenyl-1,3,5-diazaphosphorine

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$211.00
500 MG
$790.00

$211.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$211.00
500 MG
$790.00

About This Item

Empirical Formula (Hill Notation):
C23H25N2P
CAS Number:
Molecular Weight:
360.43
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$211.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: ligand

functional group

phosphine

storage temp.

−20°C

SMILES string

P2(CN(CN(C2)c4ccc(cc4)C)c3ccc(cc3)C)c1ccccc1

InChI

1S/C23H25N2P/c1-19-8-12-21(13-9-19)24-16-25(22-14-10-20(2)11-15-22)18-26(17-24)23-6-4-3-5-7-23/h3-15H,16-18H2,1-2H3

InChI key

UBTLQANULOJJIY-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service