Skip to Content
Merck
All Photos(2)

Key Documents

916242

Sigma-Aldrich

Chiralyst Ru803

Umicore

Synonym(s):

(S-MeOBIPHEP)Ru(OAc)2, Diacetato[(S)-2,2′-bis(diphenylphosphino)-6,6′-dimethoxy-1,1′-biphenyl]ruthenium(II), Ru[(S)-MeO-BIPHEP](OAc)2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$536.00

$536.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$536.00

About This Item

Empirical Formula (Hill Notation):
C42H38O6P2Ru
CAS Number:
Molecular Weight:
801.77
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$536.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

Quality Level

form

powder

mp

149-154 °C

Application

Chiralyst Ru803 is a homogeneous catalyst useful for asymmetric hydrogenation and transfer hydrogenation.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service