Skip to Content
Merck
All Photos(1)

Key Documents

91408

Supelco

BDE No 37

analytical standard

Synonym(s):

3,4,4′-TriBDE, 3,4,4′-Tribromodiphenyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$184.00

$184.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$184.00

About This Item

Empirical Formula (Hill Notation):
C12H7Br3O
CAS Number:
Molecular Weight:
406.90
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$184.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

48-52 °C

format

neat

SMILES string

Brc1c(ccc(c1)Oc2ccc(cc2)Br)Br

InChI

1S/C12H7Br3O/c13-8-1-3-9(4-2-8)16-10-5-6-11(14)12(15)7-10/h1-7H

InChI key

YALAYFVVZFORPV-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Frederiksen et al.
International journal of hygiene and environmental health, 213(4), 233-242 (2010-05-18)
Brominated flame retardants (BFRs), in particular the polybrominated diphenyl ethers (PBDEs), have been used in consumer products for many years to increase fire resistance. Recently, developmental neurotoxicity at very low levels has increased the concern about these compounds. The major
Cathrine Thomsen et al.
Molecular nutrition & food research, 52(2), 228-237 (2008-01-11)
Very high concentrations of polybrominated diphenyl ethers (PBDE) have been reported in fish from Lake Mjøsa in Norway. This study was performed to examine the serum concentrations of PBDE and hexabromocyclododecane (HBCD) in consumers of fish from this lake and
Rebeca Cruz et al.
Chemosphere, 252, 126462-126462 (2020-03-21)
Polybrominated diphenyl ethers (PBDEs) comprise a major class of brominated flame retardants and are well-known endocrine disruptors. The dietary route, through contaminated seafood consumption, is a main contributor to human exposure. Hence, the aim of this work was to provide
Miao Yu et al.
Environmental science & technology, 47(23), 13494-13501 (2013-11-07)
There is currently limited knowledge on PBDE metabolism in plants although they could play an important role in the environmental transformation of these persistent organic pollutants. In this study, pumpkin (Cucurbita maxima × C. moschata) was chosen as the model
Kamila Kalachova et al.
Talanta, 105, 109-116 (2013-04-20)
In this study, gas chromatography (GC) coupled to triple quadrupole tandem mass spectrometry (MS/MS) operated in electron ionisation mode (EI) has been shown to be an effective tool for the (ultra)trace analysis of several representative brominated flame retardants (BFRs) including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service