Skip to Content
Merck
All Photos(1)

Key Documents

90269

Supelco

Color Reference Solutions

related to Ph.Eur Color intensity testing, analytical standard, Set of colors GY

Synonym(s):

Color calibration standard greenish yellow (GY1 to GY7)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$479.00

$479.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$479.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$479.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Agency

according to Ph. Eur.

shelf life

limited shelf life, expiry date on the label

packaging

ampule of 10 mL

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

The kit contains a set of seven greenish yellow (identified sequentially from GY1 to GY7) color reference standards and a 1% hydrogen chloride solution in water. The solutions are produced from three primary metal salt solutions of ferric(III) chloride, cobalt(II) chloride, and copper(II) sulfate pentahydrate, in accordance with chapter 2.2.2 ′Degree of colouration of liquids′ of the European Pharmacopoeia (Ph. Eur.).

Application

The reference standards are used as calibration and/or quality control standards to measure the degree of coloration of liquids in the brown-yellow-red color range.

Features and Benefits

  • Ready-to-use solutions
  • Prepared and qualified in accordance with Ph. Eur.
  • Available in tamper-evident 10 mL ampules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Components

This is a set of the following color reference solutions according to PhEur:

  • Colour Reference Solution GY1 - 10mL ampule
  • Colour Reference Solution GY2 - 10mL ampule
  • Colour Reference Solution GY3 - 10mL ampule
  • Colour Reference Solution GY4 - 10mL ampule
  • Colour Reference Solution GY5 - 10mL ampule
  • Colour Reference Solution GY6 - 10mL ampule
  • Colour Reference Solution GY7 - 10mL ampule
  • Blind sample : Hydrochloric acid solution, 1% in water - 10mL ampule

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What are the empty sample vials used for the Color reference solutions acc. to Ph Eur, 90269-1SET-F?

    1 answer
    1. The recommended empty ampule vials are 72671 (Ampules 10 ml size)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service