Skip to Content
Merck
All Photos(2)

Key Documents

90139

Supelco

Schisandrin A

analytical standard

Synonym(s):

Deoxyschisandrin, Deoxyschizandrin, Schizandrin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](C1)[C@H](C)CC2=C(C3=C1C=C(OC)C(OC)=C3OC)C(OC)=C(OC)C(OC)=C2

InChI

1S/C24H32O6/c1-13-9-15-11-17(25-3)21(27-5)23(29-7)19(15)20-16(10-14(13)2)12-18(26-4)22(28-6)24(20)30-8/h11-14H,9-10H2,1-8H3/t13-,14+

InChI key

JEJFTTRHGBKKEI-OKILXGFUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Schisandrin A is a lignan, a phytonutrient found mostly in the medicinal plants belonging to the Schisandra species. Lignans are bioactive compounds that can play an important role in preventing liver injuries, carcinogenesis of the liver and lipid peroxidation.[1]

Application

Schisandrin A (Deoxyschisandrin) may be used as an analytical standard for the determination of the analyte in rat plasma[2] and fruits of Schisandra chinensis[3][4] by chromatography based techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of nine lignans using pressurized liquid extraction and HPLC-DAD in the fruits of Schisandra chinensis
Lee HJ and Kim CY
Food Chemistry, 120(4), 1224-1228 (2010)
Hui Sun et al.
Journal of separation science, 36(3), 485-491 (2013-01-11)
Shengmaisan (SMS) is a traditional Chinese medicine prescription widely used for the treatment of cardiovascular diseases in Asia. Its lignans are major components responsible for therapeutic action. A rapid and specific UPLC-Q-TOF/MS has been developed and validated for simultaneous quantification
Determination of lignans and phenolic components of Schisandra chinensis (Turcz.) Baill. using HPLC-ESI-ToF-MS and HPLC-online TEAC: Contribution of individual components to overall antioxidant activity and comparison with traditional antioxidant assays
Mocan A, et al.
Journal of functional foods, 24, 579-594 (2016)
Separation and determination of lignans from seeds of Schisandra species by micellar electrokinetic capillary chromatography using ionic liquid as modifier
Tian K, et al.
Journal of Chromatography A, 1078(1-2), 181-187 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service