Skip to Content
Merck
All Photos(1)

Key Documents

900633

Sigma-Aldrich

Dichloromethane

anhydrous, contains 40-150 ppm amylene as stabilizer, ZerO2®, ≥99.8%

Synonym(s):

Degassed and low oxygen dichloromethane, Dichloromethane ZerO2®, Methylene chloride

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 2 ML
$193.00

$193.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
4 X 2 ML
$193.00

About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21
grade:
ZerO2®
Assay:
≥99.8%
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

$193.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

grade

ZerO2®

Quality Level

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

1223 °F

quality

anhydrous

contains

40-150 ppm amylene as stabilizer

expl. lim.

22 %

impurities

≤0.001% water
water

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service