Skip to Content
Merck
All Photos(4)

Key Documents

89677

Sigma-Aldrich

Toluene

absolute, over molecular sieve (H2O ≤0.005%), ≥99.7% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12191501
eCl@ss:
39011102
PubChem Substance ID:

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

grade

absolute

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

997 °F

quality

over molecular sieve (H2O ≤0.005%)

expl. lim.

7 %

color

colorless

refractive index

n/D 1.496 (lit.)
n20/D 1.497

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

recommended

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng-Suan Cai et al.
Journal of chromatography. A, 1173(1-2), 88-97 (2007-10-30)
In this work, we evaluate the performance of liquid chromatography-atmospheric pressure photoionization-mass spectrometry (LC-APPI-MS) for non-aqueous reversed phase analysis of six triacylglycerol model compounds using six binary mobile phases including MeOH/iPrOH, MeOH/CHCl(3), MeOH/CH(2)Cl(2), CH(3)CN/iPrOH, CH(3)CN/CHCl(3), and CH(3)CN/CH(2)Cl(2). All mobile phases
V Melissinaki et al.
Biofabrication, 3(4), 045005-045005 (2011-09-21)
This study reports on the production of high-resolution 3D structures of polylactide-based materials via multi-photon polymerization and explores their use as neural tissue engineering scaffolds. To achieve this, a liquid polylactide resin was synthesized in house and rendered photocurable via
Structural study on syndiotactic polystyrene: 2. Crystal structure of molecular compound with toluene.
Chatani Y, et al.
Polymer, 34(8), 1620-1620 (1993)
T Biegert et al.
European journal of biochemistry, 238(3), 661-668 (1996-06-15)
Toluene is degraded anoxically to CO2 by the denitrifying bacterium Thauera aromatica. Toluene first becomes oxidized to benzoyl-CoA by O2-independent reactions. Benzoyl-CoA is then reduced to non-aromatic products by benzoyl-CoA reductase. We set out to study the reactions employed for
A Pseudomonas thrives in high concentrations of toluene.
Inoue A and Horikoshi K.
Nature, 338, 264-266 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service