Skip to Content
Merck
All Photos(1)

Documents

89451

Millipore

Heptakis(3-O-acetyl-2,6-di-O-methyl)-β-cyclodextrin

suitable for GC

Synonym(s):

3-O-Acetyl-2,6-di-O-methyl-β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H112O42
CAS Number:
Molecular Weight:
1625.61
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NB.21

technique(s)

gas chromatography (GC): suitable

Quality Level

mp

110-125 °C

solubility

acetone: soluble
chloroform: soluble

SMILES string

CO[C@@H]1[C@@H](OC(C)=O)[C@H](O[C@@H](O[C@@H]2COC)[C@H](OC)[C@@H](OC(C)=O)[C@@H]2O[C@@H](O[C@@H]3COC)[C@H](OC)[C@@H](OC(C)=O)[C@@H]3O[C@@H](O[C@@H]4COC)[C@H](OC)[C@@H](OC(C)=O)[C@@H]4O[C@H]5O[C@@H]6COC)[C@@H](COC)O[C@@H]1O[C@H]([C@@H](COC)O7)[C@H](OC(C)=O

InChI

1S/C70H112O42/c1-29(71)92-50-43-36(22-78-8)99-64(57(50)85-15)107-44-37(23-79-9)101-66(59(87-17)51(44)93-30(2)72)109-46-39(25-81-11)103-68(61(89-19)53(46)95-32(4)74)111-48-41(27-83-13)105-70(63(91-21)55(48)97-34(6)76)112-49-42(28-84-14)104-69(62(90-20)56(49)98-35(7)77)110-47-40(26-82-12)102-67(60(88-18)54(47)96-33(5)75)108-45-38(24-80-10)100-65(106-43)58(86-16)52(45)94-31(3)73/h36-70H,22-28H2,1-21H3/t36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50+,51+,52+,53+,54+,55+,56+,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,70-/m1/s1

InChI key

KCGPTTRGLYIRNO-DCBUKTJTSA-N

General description

Heptakis(3-O-acetyl-2,6-di-O-methyl)-β-cyclodextrin is a hydrophilic derivative of cyclodextrin (CyD). CyDs are cyclic oligosaccharides typically composed of six to eight glucose units. Heptakis(3-O-acetyl-2,6-di-O-methyl)-β-cyclodextrin finds application as a drug carrier that improves the low solubility, dissolution rate as well as the bioavailability of poorly water-soluble drugs.

Other Notes

Stationary phase for the GC separation of enantiomers

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitory effects of 2, 6-di-O-methyl-3-O-acetyl-β-cyclodextrins with various degrees of substitution of acetyl group on macrophage activation and endotoxin shock induced by lipopolysaccharide.
Motoyama K, et al.
Journal of Inclusion Phenomena and Macrocyclic Chemistry, 56(1-2), 75-79 (2006)
G. Stoev
Journal of Chromatography A, 589, 257-257 (1992)
Heptakis (2, 6-di-O-methyl-3-O-acetyl)-β-cyclodextrin: A Water-Soluble cyclodextrin derivative with low hemolytic activity.
Hirayama F, et al.
Journal of Pharmaceutical Sciences, 88(10), 970-975 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service