Skip to Content
Merck
All Photos(1)

Key Documents

89353

Supelco

Density Standard 1191 kg/m3

H&D Fitzgerald Ltd. Quality

Synonym(s):

Dimethyl phthalate, DMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1911460
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

autoignition temp.

1033 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

H&D Fitzgerald Ltd.

refractive index

n20/D 1.515 (lit.)

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum
pharmaceutical (small molecule)

format

mixture

SMILES string

COC(=O)c1ccccc1C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This density CRM is used for the calibration of all types of laboratory density meters in the test temperature range of 10 to 50 °C.

Features and Benefits

  • Manufactured and verified in an ISO 17025 accredited laboratory, as per UKAS
  • Carries an associated uncertainty of +/- 0.010 Kg/m3
  • Provided in a sealed glass ampoule of 10 mL to assure safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guy Bouchoux et al.
The journal of physical chemistry. A, 114(27), 7408-7416 (2010-06-22)
Aromatic substitution reactions of selected ionized benzenes derivatives (chlorobenzene, nitrobenzene, dimethylphtalates) and phenoxy cation using neutral methyl isocyanide as a nucleophile are shown to efficiently occur in the gas phase. Nitrilium ions are produced in high abundance during these processes.
Yi-Hung Chen et al.
Journal of hazardous materials, 192(3), 1017-1025 (2011-07-05)
The removal of dimethyl phthalate (DMP), which is a pollutant of concern in water environments, was carried out by catalytic ozonation with TiO(2)/Al(2)O(3) catalysts. The heterogeneous catalytic ozonation was an ozonation process combined with the catalytic and adsorptive properties of
Zhujian Huang et al.
Journal of hazardous materials, 246-247, 70-78 (2013-01-05)
The organic layered double hydroxides (LHDs)/TiO(2) composites with various mass ratios were prepared by the reconstruction of mixed metal oxides to photodegrade dimethyl phthalate (DMP). The physicochemical properties of the obtained products were analyzed by X-ray diffraction (XRD) spectra, X-ray
Wenxin Wang et al.
Environmental monitoring and assessment, 184(11), 6789-6798 (2011-12-08)
Atmospheric trace phthalic esters (PAEs) carried by total suspended particulates were systematically investigated. A total of 450 air samples were collected at six typical locations and three specific micro-environmental sites (a stadium with a new plastic track, new cars, and
Yiling Chen et al.
Journal of hazardous materials, 235-236, 92-100 (2012-08-14)
In this study, we demonstrate that the decomposition of dimethyl phthalate under microwave irradiation could be greatly enhanced over Fe@Fe(2)O(3) nanowires supported on activated carbon (Fe@Fe(2)O(3)/AC). The great enhanced decomposition of dimethyl phthalate could be attributed to a unique microwave

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service