89166
Legionella Supplement (Twin Pack)
suitable for microbiology
Synonym(s):
Legionella BCYE Growth Supplement
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile
Quality Level
shelf life
limited shelf life, expiry date on the label
application(s)
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Legionella spp.
General description
(per vial, sufficient for 500 ml medium)
Part A:
L-Cysteine hydrochloride 200.00 mg
Distilled water 5.00 ml
Part B:
Ferric pyrophosphate, soluble 125.00 mg
Distilled water 5.00 ml
One package contains 5 vials Part A and 5 vials Part B.
Part A:
L-Cysteine hydrochloride 200.00 mg
Distilled water 5.00 ml
Part B:
Ferric pyrophosphate, soluble 125.00 mg
Distilled water 5.00 ml
One package contains 5 vials Part A and 5 vials Part B.
Application
An enrichment supplement recommended for enhanced growth of Legionella species
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service