Skip to Content
Merck
All Photos(2)

Key Documents

88588

Millipore

Plate-Count-Agar (Agar 15 g/L)

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Plate-Count-Agar according to Buchbinder et al., Casein-peptone Dextrose Yeast Agar according to Buchbinder et al., Standard Methods Agar according to Buchbinder et al., Tryptone Glucose Yeast Extract Agar according to Buchbinder et al.

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$176.00

$176.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$176.00

About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

$176.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Agency

according to GB 4789.2-2016
according to ISO 17410:2019
according to ISO 4833-1:2013
according to ISO 4833-2:2013

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
casein enzymic hydrolysate, 5 g/L
dextrose, 1 g/L
yeast extract, 2.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

nonselective for Bacillus spp.
nonselective for Enterococcus spp.
nonselective for Escherichia coli
nonselective for Lactococcus spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

Application

Plate-Count-Agar is recommended for the plate count of microorganisms in food, dairy products, water and waste water. The formulation is compliant and recommended by APHA, FDA, GB and ISO committee.

Preparation Note

Suspend 23.5 g in 1 litre distilled water. Boil to dissolve the medium completely. Sterilise by autoclaving at 121°C for 15 min.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Kaczmarek et al.
Nanomaterials (Basel, Switzerland), 9(5) (2019-05-28)
During hyperthermia, magnetite nanoparticles placed in an AC magnetic field become a source of heat. It has been shown that in fluid suspensions, magnetic particles move freely and generate heat easily. However, in tissues of different mechanical properties, nanoparticle movement

Articles

Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service