Skip to Content
Merck
All Photos(1)

Key Documents

87187

Supelco

1-Tetradecene

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$89.80

$89.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$89.80

About This Item

Linear Formula:
CH3(CH2)11CH=CH2
CAS Number:
Molecular Weight:
196.37
Beilstein:
1701156
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$89.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

6.8 (vs air)

Assay

≥99.8% (GC)

autoignition temp.

455 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.436 (lit.)
n20/D 1.436

bp

251 °C (lit.)

mp

−13-−11 °C (lit.)

density

0.775 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

SMILES string

CCCCCCCCCCCCC=C

InChI

1S/C14H28/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3H,1,4-14H2,2H3

InChI key

HFDVRLIODXPAHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

1-Tetradecene may be used as:
  • An internal standard in the determination of fat content and fatty acid composition in food samples by gas chromatography analysis.[1]
  • A reference standard for the determination of contaminant, 1-tetradecene in Duero river source in Spain by solid-phase microextraction.[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental design applied to the determination of several contaminants in Duero River by solid-phase microextraction.
Salafranca J, et al.
Analytica Chimica Acta, 477(2), 257-267 (2003)
Determination of fat content and fatty acid composition through 1-min transesterification in the food sample; principles.
Suter B, et al.
European Food Research and Technology, 204(4), 252-258 (1997)
Lucas A Lane et al.
The journal of physical chemistry. B, 118(49), 14140-14147 (2014-08-27)
Quantum dots (QDs) offer distinct advantages over organic dyes and fluorescent proteins for biological imaging applications because of their brightness, photostability, and tunability. However, a major limitation is that single QDs emit fluorescent light in an intermittent on-and-off fashion called

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service