Skip to Content
Merck
All Photos(1)

Key Documents

87180

Sigma-Aldrich

5-(Tetradecanoylamino)fluorescein

for fluorescence, ≥95% (HPLC)

Synonym(s):

N-Tetradecanoyl-fluoresceinamine isomerI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H39NO6
CAS Number:
Molecular Weight:
557.68
MDL number:
UNSPSC Code:
12352200

grade

for fluorescence

Assay

≥95% (HPLC)

mp

185-190 °C (dec.)

suitability

suitable for fluorescence

SMILES string

CCCCCCCCCCCCCC(=O)Nc1ccc(c(c1)C(O)=O)C2=C3C=CC(=O)C=C3Oc4cc(O)ccc24

Other Notes

Fluorescent enhancer in chemiluminescent determination of serum alkaline phosphatase

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Girotti et al.
Analytical Letters, 27, 323-323 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service