Skip to Content
Merck
All Photos(1)

Key Documents

86944

Sigma-Aldrich

Hexamethyldisilazane

produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)

Synonym(s):

Silazane HMN, HMDS

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$278.00
5 KG
$1,360.00

$278.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$278.00
5 KG
$1,360.00

About This Item

Linear Formula:
(CH3)3SiNHSi(CH3)3
CAS Number:
Molecular Weight:
161.39
Beilstein:
635752
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$278.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

grade

produced by Wacker Chemie AG, Burghausen, Germany

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.407 (lit.)

bp

125 °C (lit.)

SMILES string

C[Si](C)(C)N[Si](C)(C)C

InChI

1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3

InChI key

FFUAGWLWBBFQJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethyldisilazane (HDMS) is a bulk organo silicon compound.[1][2]It finds major application as a silanizing agent, silylation with HMDS is carried out in the presence of an acid catalyst.[3]The hydrophobic nature of HDMS coatings on various nanoparticles renders them resistant to water contamination and flocculation′s, during synthesis.[2]

Application

It has been used as a
  • modifier to control the shape, formation of agglomerates,surface area and pore size of silica particles.
  • precursor for the depositing of SiNx on diamond like carbon films.
  • protective hydrophobic coating for nanoparticles.[1]

Other Notes

prices for bulk quantities on request

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

52.5 °F - closed cup

Flash Point(C)

11.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guandong Zhang et al.
Materials science & engineering. C, Materials for biological applications, 30(1), 92-97 (2010-01-01)
This paper describes the synthesis and surface engineering of core/shell-type iron/iron oxide nanoparticles for magnetic hyperthermia cancer therapy. Iron/iron oxide nanoparticles were synthesized from microemulsions of NaBH(4) and FeCl(3), followed by surface modification in which a thin hydrophobic hexamethyldisilazane layer
I Schneegass et al.
Lab on a chip, 1(1), 42-49 (2004-04-22)
Flow-through chip thermocyclers can be used in miniaturized rapid polymerase chain reaction (PCR) despite their high surface to volume ratio of samples. We demonstrated that a thermocycler made of silicon and glass chips and containing thin film transducers for heating
Surface modification of nano-sized HZSM-5 and HFER by pre-coking and silanization.
J. Catal., 251(2), 258-270 (2007)
Florian Gaertner et al.
Cell, 171(6), 1368-1382 (2017-12-02)
Blood platelets are critical for hemostasis and thrombosis and play diverse roles during immune responses. Despite these versatile tasks in mammalian biology, their skills on a cellular level are deemed limited, mainly consisting in rolling, adhesion, and aggregate formation. Here
Ashang Luwang Laiva et al.
International journal of pharmaceutics, 483(1-2), 115-123 (2015-02-15)
Electrospun hybrid nanofibrous scaffolds have gained much importance in the field of tissue engineering and drug delivery applications owing to its multifaceted properties. In this study, the properties of composite polycaprolactone (PCL)/silk fibroin (SF) nanofibrous scaffolds was investigated as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service