Skip to Content
Merck
All Photos(1)

Key Documents

86608

Supelco

Tetraethylammonium bromide

suitable for ion pair chromatography, LiChropur, ≥99.0%

Synonym(s):

TEA bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Br)
CAS Number:
Molecular Weight:
210.16
Beilstein:
3563430
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21
Assay:
≥99.0% (AT)
≥99.0%
technique(s):
ion pair chromatography: suitable

description

cationic

Quality Level

Assay

≥99.0% (AT)
≥99.0%

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

mp

285 °C (dec.) (lit.)

λ

10 % in H2O

UV absorption

λ: 240 nm Amax: 0.04
λ: 250 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Br-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

HWCKGOZZJDHMNC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The use of ion-pairing reagents as mobile phase additives allows the separation of ionic and highly polar substances on reversed phase HPLC columns. The purity of mobile phase additives is of utmost importance to their successful application. Sigma-Aldrich offers an outstanding range of tailor-made reagents for anionic (quaternary ammonium and phosphonium salts) and cationic (alkanesulfonates) determination. All mobile phase additives are subject to rigorous testing with special emphasis on the requirements of modern reversed phase HPLC:

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanomechanics of cation-π interactions in aqueous solution.
Qingye Lu et al.
Angewandte Chemie (International ed. in English), 52(14), 3944-3948 (2013-03-02)
Jayabalan Nirmal et al.
Experimental eye research, 116, 27-35 (2013-07-31)
The present study was conducted to test the hypothesis; OCT may be active from blood-to-vitreous for the uptake of its substrates. Ocular uptake of Tetraethylammonium (TEA) across blood ocular barriers and the tissue distribution was evaluated in vivo in New
Lei Yang et al.
The Journal of general physiology, 140(5), 529-540 (2012-10-31)
We investigated the effects of changing extracellular K(+) concentrations on block of the weak inward-rectifier K(+) channel Kir1.1b (ROMK2) by the three intracellular cations Mg(2+), Na(+), and TEA(+). Single-channel currents were monitored in inside-out patches made from Xenopus laevis oocytes
Ling Xiao et al.
Circulation research, 112(10), 1310-1322 (2013-03-28)
A chromosomal haplotype producing cardiac overexpression of dipeptidyl peptidase-like protein-6 (DPP6) causes familial idiopathic ventricular fibrillation. The molecular basis of transient outward current (I(to)) in Purkinje fibers (PFs) is poorly understood. We hypothesized that DPP6 contributes to PF I(to) and
P Alvarez et al.
Neuroscience, 225, 269-282 (2012-08-28)
While chronic pain is a main symptom in endometriosis, the underlying mechanisms and effective therapy remain elusive. We developed an animal model enabling the exploration of ectopic endometrium as a source of endometriosis pain. Rats were surgically implanted with autologous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service