Skip to Content
Merck
All Photos(1)

Key Documents

84480

Sigma-Aldrich

Sanguinarine chloride

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14ClNO4
CAS Number:
Molecular Weight:
367.78
Beilstein:
3832138
MDL number:
UNSPSC Code:
12352203

Assay

≥98.0% (HPLC)

solubility

methanol: 20 mg/mL, clear, orange

SMILES string

[Cl-].C[n+]1cc2c3OCOc3ccc2c4ccc5cc6OCOc6cc5c14

InChI

1S/C20H14NO4.ClH/c1-21-8-15-12(4-5-16-20(15)25-10-22-16)13-3-2-11-6-17-18(24-9-23-17)7-14(11)19(13)21;/h2-8H,9-10H2,1H3;1H/q+1;/p-1

InChI key

GIZKAXHWLRYMLE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Vlachojannis et al.
Phytotherapy research : PTR, 26(10), 1423-1426 (2012-02-10)
The rhizome of Sanguinaria canadensis (SC, bloodroot) contains an active principle with antimicrobial, antiinflammatory, antioxidative and immunomodulatory effects. For this reason SC extract has been added to toothpastes and mouthwashes in various concentrations. When tested separately, neither the toothpastes nor
Saptaparni Ghosh et al.
Biochimica et biophysica acta, 1830(8), 4189-4201 (2013-04-09)
Interaction of putative anticancer agent sanguinarine with two quadruplex forming sequences, human telomeric DNA (H24) and NHE III1 upstream of the P1 promoter of c-myc (Pu27), has been studied to understand the structural basis of the recognition. Absorption, fluorescence and
Min Ho Han et al.
Toxicology letters, 220(2), 157-166 (2013-05-11)
We examined the effects of sanguinarine, a benzophenanthridine alkaloid, on reactive oxygen species (ROS) production and the association of these effects with apoptotic cell death in a human colorectal cancer HCT-116 cell line. Sanguinarine generated ROS, which was followed by
Jia-Ying Xu et al.
Oncology reports, 28(6), 2264-2270 (2012-09-12)
Sanguinarine, a natural benzophenanthridine alkaloid, has been shown to possess anticancer activity in vitro and in vivo. In the present study, we demonstrated that sanguinarine caused a dose-dependent inhibition of growth in HeLa and SiHa human cervical cancer cells, i.e., 2.43 µmol/l (IC50)
Maidul Hossain et al.
Journal of biomolecular structure & dynamics, 30(2), 223-234 (2012-06-19)
The interaction of the natural plant alkaloid and anticancer agent sanguinarine with tRNA(phe) has been investigated by spectroscopic and calorimetric techniques. Sanguinarine iminium binds to tRNA(phe) cooperatively; alkanolamine does not bind but in presence of large tRNA(phe) concentration, a conversion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service