Skip to Content
Merck
All Photos(1)

Key Documents

83934

Sigma-Aldrich

Tris[3-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)phenyl]phosphine

technical, ≥95% (GCF)

Synonym(s):

Tris[3-(1H,1H,2H,2H-perfluorodecyl)phenyl]phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H24F51P
CAS Number:
Molecular Weight:
1600.60
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

technical

Assay

≥95% (GCF)

impurities

~5% phospinoxide (31P-NMR)

mp

90-100 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc1cccc(c1)P(c2cccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c2)c3cccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c3

Other Notes

Phosphine ligand for preparing the Bannwarth catalyst (catalyst for Stille and Suzuki coupling)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helvetica Chimica Acta, 84, 735-735 (2001)
S. Schneider et al.
Angewandte Chemie (International Edition in English), 112, 4293-4293 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service