Skip to Content
Merck
All Photos(1)

Key Documents

83714

Millipore

XLT4 Agar Supplement

suitable for microbiology

Synonym(s):

Sodium tetradecyl sulfate, 7-Ethyl-2-methyl-4-undecyl sodium salt, Niaproof® 4, Sodium tetradecyl sulfat, Sodium7-ethyl-2-methyl-4-undecyl sulfate, Tergitol 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$72.40

$72.40


Estimated to ship onMay 17, 2025


Request more information

Select a Size

Change View
100 ML
$72.40

About This Item

Empirical Formula (Hill Notation):
C14H29NaO4S
CAS Number:
Molecular Weight:
316.43
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

$72.40


Estimated to ship onMay 17, 2025


Request more information

sterility

sterile

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

suitability

Salmonella spp.
enterobacteriaceae

SMILES string

[Na+].CCCCC(CC)CCC(CC(C)C)OS([O-])(=O)=O

InChI

1S/C14H30O4S.Na/c1-5-7-8-13(6-2)9-10-14(11-12(3)4)18-19(15,16)17;/h12-14H,5-11H2,1-4H3,(H,15,16,17);/q;+1/p-1

InChI key

FVEFRICMTUKAML-UHFFFAOYSA-M

Application

A supplement for XLT4 Agar Base, for the isolation and identification of pathogenic Enterobacteriaceae, especially Salmonella spp.

Legal Information

Niaproof is a registered trademark of Niacet Corp.
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Fatimi et al.
Acta biomaterialia, 8(7), 2712-2721 (2012-04-11)
Endovascular repair of abdominal aortic aneurysms with a stent graft is limited by the persistence or recurrence of endoleaks. These are believed to be related to the recanalization of the aneurismal sac by endothelialized neochannels, which could lead to late

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service