Skip to Content
Merck
All Photos(1)

Key Documents

81268

Sigma-Aldrich

Poly(ethylene glycol)

Poly(ethylene glycol)

BioUltra, for molecular biology, 8,000

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$98.80
1 KG
$317.00
5 KG
$1,070.00

$98.80


Estimated to ship onApril 06, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$98.80
1 KG
$317.00
5 KG
$1,070.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.26

$98.80


Estimated to ship onApril 06, 2025


Request a Bulk Order

grade

for molecular biology

Quality Level

product line

BioUltra

form

powder

mol wt

Mr 7000-9000

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

mp

59-64 °C

solubility

H2O: 50 mg/mL at 20 °C, clear, colorless

density

1.0845 g/mL at 70 °C
1.0689 g/mL at 90 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.15
λ: 280 nm Amax: 0.10

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. PEG also interacts with cell membrane, thereby allowing cell fusion.[1][2]

Application

Poly(ethylene glycol) has been used as a virioplankton concentrating agent.[3][4] It has been used in the procedure involving ligation of 5′ mRNAs to RNA oligo linker.[5]

Other Notes

Precipitant in the large-scale preparation of λgt10 bacteriophage[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service