Skip to Content
Merck
All Photos(2)

Key Documents

80714

Supelco

Tanshinone I

analytical standard

Synonym(s):

Tanshinon I, Tanshinone A, Tanshinquinone I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12O3
CAS Number:
Molecular Weight:
276.29
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Cc1coc2-c3ccc4c(C)cccc4c3C(=O)C(=O)c12

InChI

1S/C18H12O3/c1-9-4-3-5-12-11(9)6-7-13-15(12)17(20)16(19)14-10(2)8-21-18(13)14/h3-8H,1-2H3

InChI key

AIGAZQPHXLWMOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tanshinone I is a bioconstituent, belonging to the class of tanshinone compounds isolated from Salvia miltiorrhiza Bunge. It is commonly used to dilate coronary arteries, increase coronary flow, modulate mutagenic activity and protect the myocardium against ischaemia.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tanshinone I may be used as an analytical reference standard for the quantification of the analyte in pharmaceutical formulations[1] and roots of Salvia miltiorrhiza[2] using different chromatography techniques.

Biochem/physiol Actions

Phenanthrenequinone constituent of Chinese medicinal herb Danshen (Salvia miltiorrhiza). Modulates or prevents breast cancer metastasis by regulating adhesion molecules. Anti-inflammatory.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of the fingerprints for the quality of the roots of Salvia miltiorrhiza and its related preparations by HPLC-DAD and LC-MSn
Liu H-A, et al.
Journal of Chromatography. B, Biomedical Applications, 846(1-2), 32-41 (2007)
Simultaneous determination of cryptotanshinone, tanshinone I and tanshinone IIA in traditional Chinese medicinal preparations containing Radix salvia miltiorrhiza by HPLC
Shi Z, et al.
Journal of Pharmaceutical and Biomedical Analysis, 37(3), 481-486 (2005)
Ying Luo et al.
PloS one, 8(2), e56774-e56774 (2013-02-26)
We previously showed that tanshinone IIA ameliorated the hypoxia-induced pulmonary hypertension (HPH) partially by attenuating pulmonary artery remodeling. The hypoxia-induced proliferation of pulmonary artery smooth muscle cells (PASMCs) is one of the major causes for pulmonary arterial remodeling, therefore the
Wen-Quan Wang et al.
Journal of hematology & oncology, 5, 69-69 (2012-11-10)
Promotion of endothelial normalization restores tumor oxygenation and obstructs tumor cells invasion, intravasation, and metastasis. We therefore investigated whether a vasoactive drug, tanshinone IIA, could inhibit metastasis by inducing vascular normalization after palliative resection (PR) of hepatocellular carcinoma (HCC). A
Qing Cai et al.
Chinese journal of integrative medicine, 18(11), 837-845 (2012-10-23)
To investigate the protective effects of sodium tanshinone B (STB) on brain damage following focal ischemia-reperfusion (I/R) injury through interfering with N-methyl-D-aspartic acid receptor (NMDAR) and excitatory and inhibitory amino acids, and evaluate the potential mechanisms of the neuroprotective activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service