Skip to Content
Merck
All Photos(2)

Key Documents

79588

Supelco

D-(−)-Salicin

analytical standard

Synonym(s):

2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, Salicyl alcohol glucoside, Saligenin β-D-glucoside

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$346.00

$346.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$346.00

About This Item

Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
Beilstein:
89593
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$346.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC[C@H]1O[C@@H](Oc2ccccc2CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C13H18O7/c14-5-7-3-1-2-4-8(7)19-13-12(18)11(17)10(16)9(6-15)20-13/h1-4,9-18H,5-6H2/t9-,10-,11+,12-,13-/m1/s1

InChI key

NGFMICBWJRZIBI-UJPOAAIJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hanna Oltean et al.
The Cochrane database of systematic reviews, 12(12), CD004504-CD004504 (2014-12-24)
Low-back pain (LBP) is a common condition and imposes a substantial economic burden upon people living in industrialized societies. A large proportion of people with chronic LBP use complementary and alternative medicine (CAM), visit CAM practitioners, or both. Several herbal
Ken Keefover-Ring et al.
PloS one, 9(10), e107189-e107189 (2014-10-10)
Salicinoids are well-known defense compounds in salicaceous trees and careful screening at the population level is warranted to fully understand their diversity and function. European aspen, Populus tremula, is a foundation species in Eurasia and highly polymorphic in Sweden. We
Mahla Mirali et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 969, 149-161 (2014-09-01)
A systematic set of optimization experiments was conducted to design an efficient extraction and analysis protocol for screening six different sub-classes of phenolic compounds in the seed coat of various lentil (Lens culinaris Medik.) genotypes. Different compounds from anthocyanidins, flavan-3-ols
Jasbir D Upadhyaya et al.
PloS one, 9(10), e110373-e110373 (2014-10-24)
Activation of bitter taste receptors (T2Rs) in human airway smooth muscle cells leads to muscle relaxation and bronchodilation. This finding led to our hypothesis that T2Rs are expressed in human pulmonary artery smooth muscle cells and might be involved in
Véronique Gilard et al.
Journal of pharmaceutical and biomedical analysis, 102, 476-493 (2014-12-03)
One hundred and fifty dietary supplements (DS) marketed to increase sexual performance were analyzed. All these formulations were claimed to contain only natural compounds, plant extracts and/or vitamins. (1)H NMR spectroscopy was used for detecting the presence of adulterants and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service