Skip to Content
Merck
All Photos(1)

Key Documents

79332

Millipore

Davis Minimal Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Minimal Agar, Davis

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$235.00

$235.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$235.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$235.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
ammonium sulfate, 1 g/L
dextrose, 1 g/L
dipotassium phosphate, 7 g/L
magnesium sulfate, 0.1 g/L
monopotassium phosphate, 2 g/L
sodium citrate, 0.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

food and beverages
life science and biopharma

microbiology

suitability

nonselective for Escherichia coli
nonselective for coliforms

Application

for the isolation and characterization of nutrional mutants of Escherichia coli

Preparation Note

Suspend 26.6 g in 1000 ml of distilled water. Bring to the boil and dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes. Mix well and pour into sterile petri dishes or dispense as desired.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald M. Atlas
Handbook of Microbiological Media (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service