Skip to Content
Merck
All Photos(1)

Key Documents

79270

Sigma-Aldrich

1-Phenyl-1-trimethylsiloxyethylene

purum, ≥98.0% (GC)

Synonym(s):

α-(Trimethylsiloxy)styrene, Acetophenone enol trimethylsilyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$25.30
25 MG
$33.35
50 MG
$48.30
100 MG
$74.75
250 MG
$147.20
1 G
$370.30
5 G
$1,322.50

Vendor SKU: A302163-10MG

$25.30


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
10 MG
$25.30
25 MG
$33.35
50 MG
$48.30
100 MG
$74.75
250 MG
$147.20
1 G
$370.30
5 G
$1,322.50

About This Item

Linear Formula:
C6H5C(=CH2)OSi(CH3)3
CAS Number:
Molecular Weight:
192.33
Beilstein:
1306914
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Vendor SKU: A302163-10MG

$25.30


Ships in 7 days - from Aldrich Partner.

vapor density

>1 (vs air)

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.502 (lit.)
n20/D 1.503

bp

88-89 °C/11 mmHg (lit.)

density

0.938 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[Si](C)(C)OC(=C)c1ccccc1

InChI

1S/C11H16OSi/c1-10(12-13(2,3)4)11-8-6-5-7-9-11/h5-9H,1H2,2-4H3

Looking for similar products? Visit Product Comparison Guide

Other Notes

Versatile intermediate[1][2][3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.H. Mashraqui et al.
The Journal of Organic Chemistry, 49, 2513-2513 (1984)
J.K. Rasmussen
Synthesis, 91-91 (1977)
I. Fleming
Chimia, 34, 265-265 (1980)
E.W. Colvin
Chemical Society Reviews, 7, 15-15 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service