Skip to Content
Merck
All Photos(1)

Key Documents

78855

Supelco

Redox standard

465 mV

Synonym(s):

Redox Potential Standard

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$255.00

$255.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$255.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$255.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

description

redox potential: 465 mV ±5 mV, 25°C

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
pharmaceutical

format

mixture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This Redox standard has a potential of 465 mV ±5 mV. It refers to the potential of the platinum electrode versus the Ag/AgCl reference electrode in 3M potassium chloride.

Application

Redox standard acts as a control standard rather than a calibration standard which controls the functionality of the sensing and reference electrodes.

Features and Benefits

  • Broad range of values
  • Very high specifications (±5mV)
  • Detailed safety data sheet (SDS) available
  • Certified with proven verifiable accuracy and uncertainty of measurement
  • Guaranteed stability throughout its entire shelf life

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Il prodotto è certificato ISO 17034?

    1 answer
    1. This is not a Certified Reference Material but an analytical standard therefore it is not ISO 17034 certified.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service