Skip to Content
Merck
All Photos(3)

Key Documents

77699

Sigma-Aldrich

2-Phenoxyethanol

≥99%

Synonym(s):

Ethylene glycol monophenyl ether, Phenylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1364011
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
viscous liquid
Assay:
≥99%

Quality Level

Assay

≥99%

form

viscous liquid

refractive index

n20/D 1.539

bp

244-246 °C

mp

11-13 °C

solubility

soluble, clear

density

1.107 g/mL at 20 °C (lit.)

functional group

hydroxyl
phenoxy

SMILES string

OCCOc1ccccc1

InChI

1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

QCDWFXQBSFUVSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Phenoxyethanol is used as a fish anesthetic agent.

Application

2-Phenoxyethanol may be used as a starting material to synthesize diorganodiselenides derivatives. It can undergo aromatic alkylation with fluoren-9-one to form 9,9-bis[4-(2-hydroxyethoxy)phenyl]fluorene in the presence of Ti4+ cation-exchanged montmorillonite catalyst.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficacy of 2-phenoxyethanol as an anaesthetic for adult redline torpedo fish, Sahyadria denisonii (day 1865).
Varkey T
International Journal of Zoology (2014)
Mehmet Çağrı Akgündüz et al.
Scientific reports, 10(1), 1209-1209 (2020-01-29)
In this study, the toxic effects of phenoxyethanol (Phy-Et), which is widely used in cosmetic industry, has been investigated with Allium test by means of physiological, cytogenetic, anatomical and biochemical parameters. To determine the changes in physiological reactions weight gain
Anaesthetic qualities of eugenol and 2-phenoxyethanol and their effect on some haematological parameters in farmed European sea bass (Dicentrarchus labrax L.).
Filiciotto F
Journal of Animal and Veterinary Advances, 11, 494-502 (2012)
Simple and clean synthesis of 9, 9-bis [4-(2-hydroxyethoxy) phenyl] fluorene from the aromatic alkylation of phenoxyethanol with fluoren-9-one catalysed by titanium cation-exchanged montmorillonite.
Ebitani K
Green Chemistry, 2(4), 157-160 (2000)
Catarina C V Oliveira et al.
International journal of molecular sciences, 21(23) (2020-12-03)
Kisspeptin regulates puberty and reproduction onset, acting upstream of the brain-pituitary-gonad (HPG) axis. This study aimed to test a kisspeptin-based hormonal therapy on cultured Senegalese sole (G1) breeders, known to have reproductive dysfunctions. A single intramuscular injection of KISS2-10 decapeptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service