Skip to Content
Merck
All Photos(1)

Documents

77505

Sigma-Aldrich

Potassium iodide

purum p.a., ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor pressure

1 mmHg ( 745 °C)

Quality Level

grade

purum p.a.

Assay

≥99.0% (AT)

form

solid

pH

~6.9 (20 °C, 50 g/L)

mp

681 °C (lit.)

solubility

water: soluble(lit.)

anion traces

bromide, chloride (as Cl-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Na: ≤2000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 889-889 (1994)
H B Li et al.
Journal of chromatography. A, 918(2), 335-339 (2001-06-16)
A novel method for the determination of iodide by size exclusion chromatography was established. The method was simple and highly sensitive with good precision. Iodide was converted to iodine, then sequestered with starch, and separated from the matrix using a
V Witko-Sarsat et al.
Journal of immunology (Baltimore, Md. : 1950), 161(5), 2524-2532 (1998-09-02)
We previously demonstrated the presence of advanced oxidation protein products (AOPP), a novel marker of oxidative stress in the plasma of uremic patients receiving maintenance dialysis. The present study in a cohort of 162 uremic patients showed that plasma concentrations
Alumina-supported potassium iodide as a heterogeneous catalyst for biodiesel production from soybean oil.
Xie W and Li H.
J. Mol. Catal. A: Chem., 255(1), 1-9 (2006)
S C Clement et al.
Pediatric blood & cancer, 60(11), 1833-1838 (2013-07-09)
Thyroid dysfunction has been reported in up to 52% of patients 1.4 years after treatment with (131) I-Metaiodobenzylguanidine (MIBG) in children with neuroblastoma (NBL), despite the use of potassium-iodide (KI). Our aim was to investigate if the incidence and severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service