Skip to Content
Merck
All Photos(1)

Key Documents

74850

Sigma-Aldrich

1-Octanol

puriss., ≥99.5% (GC)

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

grade

puriss.

Assay

≥99.5% (GC)

autoignition temp.

523 °F

expl. lim.

8 % (lit.)

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 744-744 (1994)
Mechanisms of PVDF membrane formation by immersion-precipitation in soft (1-octanol) and harsh (water) nonsolvents.
Young TH, et al.
Polymer, 40(19), 5315-5323 (1999)
Photocatalytic degradation of 1-octanol on anchored titanium oxide and on TiO2 powder catalysts.
Yamashita H, et al.
J. Catal., 158(1), 97-101 (1996)
Characterization of titanium-silicon binary oxide catalysts prepared by the sol-gel method and their photocatalytic reactivity for the liquid-phase oxidation of 1-octanol.
Yamashita H, et al.
The Journal of Physical Chemistry B, 102(30), 5870-5875 (1998)
John A Arnott et al.
Expert opinion on drug discovery, 7(10), 863-875 (2012-09-21)
The role of lipophilicity in drug discovery and design is a critical one. Lipophilicity is a key physicochemical property that plays a crucial role in determining ADMET (absorption, distribution, metabolism, excretion, and toxicity) properties and the overall suitability of drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service