Skip to Content
Merck
All Photos(2)

Key Documents

74748

Supelco

Patent Blue V calcium salt

analytical standard

Synonym(s):

Acid Blue 3 calcium salt, Acid Blue V calcium salt, Food Blue 5, Patent Blue V hemicalcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$217.00

$217.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$217.00

About This Item

Empirical Formula (Hill Notation):
C27H31N2O7S2 · 0.5Ca
CAS Number:
Molecular Weight:
579.71
Colour Index Number:
42051
Beilstein:
4108121
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E131
NACRES:
NA.24

$217.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Ca++].CCN(CC)c1ccc(cc1)\C(=C2/C=C\C(C=C2)=[N+](\CC)CC)c3cc(O)c(cc3S([O-])(=O)=O)S([O-])(=O)=O.CCN(CC)c4ccc(cc4)C(=C5/C=C\C(C=C5)=[N+](\CC)CC)\c6cc(O)c(cc6S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/2C27H32N2O7S2.Ca/c2*1-5-28(6-2)21-13-9-19(10-14-21)27(20-11-15-22(16-12-20)29(7-3)8-4)23-17-24(30)26(38(34,35)36)18-25(23)37(31,32)33;/h2*9-18H,5-8H2,1-4H3,(H2-,30,31,32,33,34,35,36);/q;;+2/p-2

InChI key

RTMBGDBBDQKNNZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Literature for Patent Blue Violet discusses two dyes which are very similar, Color Index Numbers 42045 and 42051. Structurally they differ with regard to the substituents on one of the aromatic rings. C.I. 42045 is 2,4-disulfo-substituted while C.I. 42051 is 2,4-disulfo-5-hydroxy-substituted. This is the latter form.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Meng et al.
Journal of virology, 93(17) (2019-06-07)
Hand, foot, and mouth disease (HFMD), a highly contagious disease in children, is caused by human enteroviruses, including enterovirus 71 (EV71), coxsackievirus A16 (CVA16), and coxsackievirus A6 (CVA6). Although HFMD is usually mild and self-limiting, EV71 infection occasionally leads to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service